• USA Home
  • 86842 - Tetrabutylammonium phosphate monobasic solution

EMAIL THIS PAGE TO A FRIEND
86842 Sigma-Aldrich

Tetrabutylammonium phosphate monobasic solution

puriss., ~1 M in H2O

Synonym: Tetrabutylammonium dihydrogen phosphate

  • CAS Number 5574-97-0

  • Linear Formula (CH3CH2CH2CH2)4N[OP(OH)2O]

  • Molecular Weight 339.45

  •  Beilstein Registry Number 5196532

  •  MDL number MFCD00064526

  •  PubChem Substance ID 329769493

Purchase

Properties

Related Categories Ammonium Salts, Chemical Synthesis, Phase Transfer Catalysts, Synthetic Reagents
InChI Key   ARRNBPCNZJXHRJ-UHFFFAOYSA-M
grade   puriss.
concentration   ~1 M in H2O
density   1.04 g/mL at 20 °C

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?