• USA Home
  • 86869 - Tetrabutylammonium tetrafluoroborate

EMAIL THIS PAGE TO A FRIEND
86869 Sigma-Aldrich

Tetrabutylammonium tetrafluoroborate

puriss., ≥99.0% (T)

  • CAS Number 429-42-5

  • Linear Formula (CH3CH2CH2CH2)4N(BF4)

  • Molecular Weight 329.27

  •  Beilstein Registry Number 3577508

  •  EC Number 207-058-8

  •  MDL number MFCD00011634

  •  PubChem Substance ID 329769329

Purchase

Properties

Related Categories Ammonium Salts, Chemical Synthesis, Phase Transfer Catalysts, Synthetic Reagents
InChI Key   NNZZSJSQYOFZAM-UHFFFAOYSA-N
grade   puriss.
assay   ≥99.0% (T)
mp   155-161 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
>199.4 °F
Flash Point(C) 
>93 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

217964 Tetrabutylammonium tetrafluoroborate, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?