• USA Home
  • 86871 - Tetrabutylammonium cyanide

EMAIL THIS PAGE TO A FRIEND
86871 Sigma-Aldrich

Tetrabutylammonium cyanide

purum, ≥95.0% (AT)

  • CAS Number 10442-39-4

  • Linear Formula (CH3CH2CH2CH2)4N(CN)

  • Molecular Weight 268.48

  •  Beilstein Registry Number 3598808

  •  EC Number 233-926-0

  •  MDL number MFCD00043221

  •  PubChem Substance ID 329769330

Purchase

Properties

Related Categories Ammonium Salts, Chemical Synthesis, Phase Transfer Catalysts, Synthetic Reagents
InChI Key   KRRBFUJMQBDDPR-UHFFFAOYSA-N
grade   purum
assay   ≥95.0% (AT)
mp   89-92 °C(lit.)

Description

Other Notes

Reagent for the synthesis of nitriles; Preparation of the complex with CuCN and use as coupling reagent; Mild reagent for transesterifications

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Supplemental Hazard Statements 
Contact with acids liberates very toxic gas.
RIDADR 
UN2811 - class 6.1 - PG 2 - MP - EHS - Toxic solids, organic, n.o.s., HI: all
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

358665 Tetrabutylammonium cyanide, 95%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?