• USA Home
  • 86875 - Tetrabutylammonium bisulfate

EMAIL THIS PAGE TO A FRIEND
86875 Sigma-Aldrich

Tetrabutylammonium bisulfate

purum, ≥97.0% (T)

Synonym: Tetrabutylammonium hydrogen sulfate

  • CAS Number 32503-27-8

  • Linear Formula (CH3CH2CH2CH2)4N(HSO4)

  • Molecular Weight 339.53

  •  Beilstein Registry Number 3599663

  •  EC Number 251-068-5

  •  MDL number MFCD00011637

  •  PubChem Substance ID 329769333

Purchase

Properties

Related Categories Ammonium Salts, Chemical Synthesis, Essential Chemicals, Phase Transfer Catalysts, Research Essentials,
InChI Key   SHFJWMWCIHQNCP-UHFFFAOYSA-M
grade   purum
assay   ≥97.0% (T)
mp   167-173 °C

Description

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

155837 Tetrabutylammonium hydrogensulfate, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?