• USA Home
  • 86883 - Tetrabutylammonium nitrate

EMAIL THIS PAGE TO A FRIEND
86883 Sigma-Aldrich

Tetrabutylammonium nitrate

purum, ≥97.0% (NT)

  • CAS Number 1941-27-1

  • Linear Formula (CH3CH2CH2CH2)4N(NO3)

  • Molecular Weight 304.47

  •  Beilstein Registry Number 3921016

  •  EC Number 217-726-0

  •  MDL number MFCD00043202

  •  PubChem Substance ID 24888802

Purchase

Properties

Related Categories Ammonium Salts, Chemical Synthesis, Phase Transfer Catalysts, Synthetic Reagents
InChI Key   QHOKENWFMZXSEU-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (NT)
mp   116-118 °C(lit.)
  116-120 °C

Description

Other Notes

Recommended as supporting electrolyte in liquid ammonia; Synthesis of nitrates from triflates; used in the inversion of configuration of alcohols via tosylates; conversion of halides into ketones

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1479 5.1 / PGII
WGK Germany 
3
RTECS 
BS5475000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

359173 Tetrabutylammonium nitrate, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?