• USA Home
  • 86886 - Tetrabutylammonium thiocyanate

EMAIL THIS PAGE TO A FRIEND
86886 Sigma-Aldrich

Tetrabutylammonium thiocyanate

purum, ≥99.0% (AT)

Synonym: Tetrabutylammonium rhodanide

  • CAS Number 3674-54-2

  • Linear Formula (CH3CH2CH2CH2)4N(SCN)

  • Molecular Weight 300.55

  •  Beilstein Registry Number 3919896

  •  EC Number 222-943-9

  •  MDL number MFCD00012140

  •  PubChem Substance ID 24888805

Purchase

Properties

Related Categories Ammonium Salts, Chemical Synthesis, Phase Transfer Catalysts, Synthetic Reagents
InChI Key   RSHBFZCIFFBTEW-UHFFFAOYSA-M
grade   purum
assay   ≥99.0% (AT)
mp   120-123 °C(lit.)
  120-125 °C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Supplemental Hazard Statements 
Contact with acids liberates very toxic gas.
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

323314 Tetrabutylammonium thiocyanate, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?