• USA Home
  • 86887 - Tetrabutylammonium p-toluenesulfonate

EMAIL THIS PAGE TO A FRIEND
86887 Sigma-Aldrich

Tetrabutylammonium p-toluenesulfonate

purum, ≥99.0% (T)

Synonym: p-Toluenesulfonic acid tetrabutylammonium salt

  • CAS Number 7182-86-7

  • Linear Formula (CH3CH2CH2CH2)4N(CH3C6H4SO3)

  • Molecular Weight 413.66

  •  Beilstein Registry Number 3643278

  •  EC Number 230-548-8

  •  MDL number MFCD00043227

  •  PubChem Substance ID 24888806

Purchase

Properties

Related Categories Ammonium Salts, Chemical Synthesis, Phase Transfer Catalysts, Synthetic Reagents
InChI Key   REAVCZWUMGIGSW-UHFFFAOYSA-M
grade   purum
assay   ≥99.0% (T)
mp   70-72 °C(lit.)

Description

Application

Supporting electrolyte

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

358681 Tetrabutylammonium p-toluenesulfonate, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?