• USA Home
  • 86894 - Tetrabutylammonium iodide

EMAIL THIS PAGE TO A FRIEND
86894 Sigma-Aldrich

Tetrabutylammonium iodide

purum, ≥98.0% (AT)

Synonym: TBAI

  • CAS Number 311-28-4

  • Linear Formula (CH3CH2CH2CH2)4N(I)

  • Molecular Weight 369.37

  •  Beilstein Registry Number 3916152

  •  EC Number 206-220-5

  •  MDL number MFCD00011636

  •  PubChem Substance ID 329769342

Purchase

Properties

Related Categories Ammonium Salts, Chemical Synthesis, Phase Transfer Catalysts, Synthetic Reagents
InChI Key   DPKBAXPHAYBPRL-UHFFFAOYSA-M
grade   purum
assay   ≥98.0% (AT)
mp   141-143 °C(lit.)
  143-147 °C

Description

Other Notes

This product has been replaced by 140775-ALDRICH | Tetrabutylammonium iodide reagent grade, 98%

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
BS5450000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?