• USA Home
  • 86901 - Tetrabutylammonium tetraphenylborate

EMAIL THIS PAGE TO A FRIEND
86901 Sigma-Aldrich

Tetrabutylammonium tetraphenylborate

puriss., ≥99.0% (NT)

Synonym: Tetraphenylboron tetrabutylammonium

  • CAS Number 15522-59-5

  • Linear Formula (CH3CH2CH2CH2)4N[B(C6H5)4]

  • Molecular Weight 561.69

  •  Beilstein Registry Number 3857215

  •  EC Number 239-562-9

  •  MDL number MFCD00011749

Purchase

Properties

Related Categories Ammonium Salts, Chemical Synthesis, Phase Transfer Catalysts, Synthetic Reagents
InChI Key   ZHCCBGAUZWZGQV-UHFFFAOYSA-N
grade   puriss.
assay   ≥99.0% (NT)
mp   230-236 °C(lit.)
  230-236 °C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

281034 Tetrabutylammonium tetraphenylborate, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?