• USA Home
  • 86902 - Tetrabutylammonium tribromide

EMAIL THIS PAGE TO A FRIEND
86902 Sigma-Aldrich

Tetrabutylammonium tribromide

purum, ≥98.0% (RT)

Synonym: TBABr3, Tetrabutylammonium bromide perbromide

  • CAS Number 38932-80-8

  • Linear Formula (CH3CH2CH2CH2)4NBr3

  • Molecular Weight 482.18

  •  Beilstein Registry Number 3746114

  •  MDL number MFCD00012110

  •  PubChem Substance ID 24888816

Purchase

Properties

Related Categories Ammonium, Ammonium Salts, Chemical Synthesis, Ionic Liquids, Phase Transfer Catalysts,
InChI Key   XXSLZJZUSYNITM-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (RT)
mp   71-76 °C(lit.)
  71-76 °C

Description

Other Notes

Brominating agent; Hofmann degradation of amides to ureas and carbamates

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?