• USA Home
  • 87029 - Multielement Ion Chromatography Anion Standard solution

EMAIL THIS PAGE TO A FRIEND
87029 Sigma-Aldrich

Multielement Ion Chromatography Anion Standard solution

analytical standard

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Anion Standards, Chromatography,
grade   analytical standard
shelf life   limited shelf life, expiry date on the label
composition   Br, 20 mg/kg
  Cl, 10 mg/kg
  F, 3 mg/kg
  NO3, 20 mg/kg
  PO4, 30 mg/kg
  SO4, 20 mg/kg
application(s)   ion chromatography: suitable
format   multi-component solution

Description

General description

filtered through a 0.45 μm membrane filter

Preparation Note

prepared with high purity salts

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?