EMAIL THIS PAGE TO A FRIEND
87160 Sigma-Aldrich

1-Tetradecanol

purum, ≥95.0% (GC)

Synonym: Myristyl alcohol, Tetradecyl alcohol

  • CAS Number 112-72-1

  • Linear Formula CH3(CH2)13OH

  • Molecular Weight 214.39

  •  Beilstein Registry Number 1742652

  •  EC Number 204-000-3

  •  MDL number MFCD00004757

Purchase

Properties

Related Categories Alcohols, Biochemicals and Reagents, Building Blocks, C11 to C30+, Chemical Synthesis,
vapor density   7.4 (vs air)
InChI Key   HLZKNKRTKFSKGZ-UHFFFAOYSA-N
grade   purum
assay   ≥95.0% (GC)
autoignition temp.   489 °F
bp   289 °C(lit.)
mp   35-39 °C(lit.)
  35-39 °C
density   0.823 g/mL at 25 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
nwg
RTECS 
XB8655000
Flash Point(F) 
298.4 °F
Flash Point(C) 
148 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?