• USA Home
  • 87293 - Tetraheptylammonium tetraphenylborate

EMAIL THIS PAGE TO A FRIEND
87293 Sigma-Aldrich

Tetraheptylammonium tetraphenylborate

Selectophore

Synonym: Tetraphenylboron tetraheptylammonium

  • CAS Number 22560-28-7

  • Empirical Formula (Hill Notation) C52H80BN

  • Molecular Weight 730.01

  •  Beilstein Registry Number 4121817

  •  MDL number MFCD00210097

  •  PubChem Substance ID 329769519

Purchase

Properties

Related Categories Additives for ISE, Analytical/Chromatography, Ion Sensor Materials More...
grade   Selectophore
InChI Key   FYJAQRCBXXYPGC-UHFFFAOYSA-N
mp   142-143 °C

Description

General description

Visit our Sensor Applications portal to learn more.

Other Notes

Lipophilic additive for NH4+-selective polymer membrane electrodes.; Lipophilic supporting electrolyte

Legal Information

Selectophore is a trademark of Sigma-Aldrich Chemie GmbH

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?