• USA Home
  • 87294 - Tetrahexadecylammonium bromide

EMAIL THIS PAGE TO A FRIEND
87294 Sigma-Aldrich

Tetrahexadecylammonium bromide

Selectophore

  • CAS Number 139653-55-7

  • Linear Formula [CH3(CH2)15]4N(Br)

  • Molecular Weight 995.64

  •  Beilstein Registry Number 5845950

  •  MDL number MFCD00075496

  •  PubChem Substance ID 329769520

Purchase

Properties

Related Categories Analytical/Chromatography, Ion Sensor Materials, Ionophores Potentiometric for ISE More...
grade   Selectophore
InChI Key   RBIRNUUGKIFHKK-UHFFFAOYSA-M
mp   97-100 °C(lit.)
  99-101 °C

Description

General description

Visit our Sensor Applications portal to learn more.

Legal Information

Selectophore is a trademark of Sigma-Aldrich Chemie GmbH

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

87298 Tetrahexadecylammonium bromide, purum, ≥98.0% (NT)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?