• USA Home
  • 87298 - Tetrahexadecylammonium bromide

EMAIL THIS PAGE TO A FRIEND
87298 Sigma-Aldrich

Tetrahexadecylammonium bromide

purum, ≥98.0% (NT)

  • CAS Number 139653-55-7

  • Linear Formula [CH3(CH2)15]4N(Br)

  • Molecular Weight 995.64

  •  Beilstein Registry Number 5845950

  •  MDL number MFCD00075496

Purchase

Properties

Related Categories Ammonium, Ammonium Salts, Chemical Synthesis, Ionic Liquids, Phase Transfer Catalysts,
InChI Key   RBIRNUUGKIFHKK-UHFFFAOYSA-M
grade   purum
assay   ≥98.0% (NT)
mp   97-100 °C(lit.)
  97-100 °C

Description

Application

Extremely lipophilic quaternary ammonium bromide

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

367524 Tetrahexadecylammonium bromide, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?