• USA Home
  • 87313 - Tetrahexylammonium dihydrogen phosphate solution

EMAIL THIS PAGE TO A FRIEND
87313 Sigma-Aldrich

Tetrahexylammonium dihydrogen phosphate solution

concentrate, for ion pair chromatography

Purchase

Properties

Related Categories Acidic Ion Pair Reagents (Anionic), Analytical Reagents, Analytical/Chromatography, Chromatography Reagents & Solvents, Ion Pair Reagents More...
grade   for ion pair chromatography
InChI Key   ITLADWLZFFSZKW-UHFFFAOYSA-M
quality   ampule
λ   in concentrate
UV absorption   λ: 210 nm Amax: 0.10
  λ: 230 nm Amax: 0.06
  λ: 330 nm Amax: 0.02
  λ: 500 nm Amax: 0.02
suitability   corresponds to standard for filter test

Description

Packaging

concentrate in ampules for 6x1 l 0.005 M ready-to-use solution

Price and Availability


All labs need water
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?