EMAIL THIS PAGE TO A FRIEND
87362 Sigma-Aldrich

Tetrahydrofuran

for luminescence, ≥99.5% (GC)

  • CAS Number 109-99-9

  • Empirical Formula (Hill Notation) C4H8O

  • Molecular Weight 72.11

  •  Beilstein Registry Number 102391

  •  EC Number 203-726-8

  •  MDL number MFCD00005356

  •  PubChem Substance ID 329769578

Purchase

Properties

Related Categories Biochemicals and Reagents, Luminescent Compounds/Detection, Solvents, Buffers and Reagents for Luminescence More...
grade   for luminescence
vapor density   2.5 (vs air)
vapor pressure   114 mmHg ( 15 °C)
  143 mmHg ( 20 °C)
InChI Key   WYURNTSHIVDZCO-UHFFFAOYSA-N
assay   ≥99.5% (GC)
autoignition temp.   610 °F
expl. lim.   11.8 %
refractive index   n20/D 1.407(lit.)
  n20/D 1.407
bp   65-67 °C(lit.)
mp   −108 °C(lit.)
solubility   H2O: soluble
density   0.889 g/mL at 25 °C(lit.)
λ   1 cm path, H2O reference
UV absorption   λ: 240 nm Amax: 0.3
  λ: 250 nm Amax: 0.2
  λ: 260 nm Amax: 0.1
  λ: 280 nm Amax: 0.03
  λ: 310 nm Amax: 0.003
suitability   in accordance for luminescence

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Supplemental Hazard Statements 
May form explosive peroxides.
RIDADR 
UN 2056 3 / PGII
WGK Germany 
1
RTECS 
LU5950000
Flash Point(F) 
1.4 °F
Flash Point(C) 
-17 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

recommended

Product #

Description

Add to Cart

Z683299 XPell indicating pellets for peroxides prevention, CE for organic solvents

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?