EMAIL THIS PAGE TO A FRIEND
87363 Sigma-Aldrich

Tetrahydrofuran

for amino acid analysis, without stabilizer

  • CAS Number 109-99-9

  • Empirical Formula (Hill Notation) C4H8O

  • Molecular Weight 72.11

  •  Beilstein Registry Number 102391

  •  EC Number 203-726-8

  •  MDL number MFCD00005356

Purchase

Properties

Related Categories Amino Acid Analysis, Amino Acid Analysis Reagents, Biochemicals and Reagents, Peptide Analysis and Characterization, Peptides More...
grade   for amino acid analysis
vapor density   2.5 (vs air)
vapor pressure   114 mmHg ( 15 °C)
  143 mmHg ( 20 °C)
InChI Key   WYURNTSHIVDZCO-UHFFFAOYSA-N
autoignition temp.   610 °F
quality   without stabilizer
expl. lim.   11.8 %
refractive index   n20/D 1.407(lit.)
bp   65-67 °C(lit.)
mp   −108 °C(lit.)
solubility   H2O: soluble
density   0.889 g/mL at 25 °C(lit.)
storage temp.   2-8°C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Application

Mobile-phase component in the HPLC analysis of triglycerides.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Supplemental Hazard Statements 
May form explosive peroxides.
RIDADR 
UN 2056 3 / PGII
WGK Germany 
1
RTECS 
LU5950000
Flash Point(F) 
1.4 °F
Flash Point(C) 
-17 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

recommended

Product #

Description

Add to Cart

Z683299 XPell indicating pellets for peroxides prevention, CE for organic solvents

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?