• USA Home
  • 87471 - Micro particles based on melamine resin, rhodamine B-marked

EMAIL THIS PAGE TO A FRIEND
87471 Sigma-Aldrich

Micro particles based on melamine resin, rhodamine B-marked

analytical standard, size: 23 μm

Synonym: Melamine-formaldehyde resin

Purchase

Properties

Related Categories Biochemicals and Reagents, Fluorescent Microparticles, Micro particles based on melamine resin, rhodamine B labeled, Microparticles
grade   analytical standard
form   aqueous suspension
  particles
concentration   2.5% (solids)
particle size   23 μm std dev <3 μm
fluorescence   λex 540 nm; λem 584
format   neat

Description

Frequently Asked Questions

Frequently Asked Questions are available for this Product.

Analysis Note

For every lot, exact values of particle size and standard deviation are determined with an accuracy of 0.01 μm using a Coulter Multisizer.

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Articles

Fluorescent Microparticles and Nanobeads

Microparticles have been found to have widespread applications in medicine, biochemistry, colloid chemistry, and aerosol research. Uses include chromatographic separation media, supports for immobili...
BioFiles 2007, 2.5, 25.
Keywords: Agglutination, Biochemistry, Catalysis, Derivatizations, Electronics, Environmental, Flow cytometry, Immobilization, Ligands, Microscopy, Nanotechnology, Separation, Solvents

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?