• USA Home
  • 87485 - Tetrahydro-4H-thiopyran-4-one

EMAIL THIS PAGE TO A FRIEND
87485 Sigma-Aldrich

Tetrahydro-4H-thiopyran-4-one

purum, ≥99.0% (GC)

  • CAS Number 1072-72-6

  • Empirical Formula (Hill Notation) C5H8OS

  • Molecular Weight 116.18

  •  Beilstein Registry Number 106464

  •  EC Number 214-015-7

  •  MDL number MFCD00006660

  •  PubChem Substance ID 329769584

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Heterocyclic Building Blocks, Other Heterocyclic Building Blocks, S-Containing More...
InChI Key   OVRJVKCZJCNSOW-UHFFFAOYSA-N
grade   purum
assay   ≥99.0% (GC)
mp   60-64 °C(lit.)
  60-64 °C

Description

Other Notes

For the preparation of 4-methoxy-3,6-dihydro-2H-thiopyran, a new protecting reagent for alcohols

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

155160 Tetrahydro-4H-thiopyran-4-one, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?