• USA Home
  • 87620 - Tetrakis(hydroxymethyl)phosphonium chloride solution

EMAIL THIS PAGE TO A FRIEND
87620 Sigma-Aldrich

Tetrakis(hydroxymethyl)phosphonium chloride solution

technical, ~80% in H2O

  • CAS Number 124-64-1

  • Linear Formula (HOCH2)4PCl

  • Molecular Weight 190.56

  •  Beilstein Registry Number 3631669

  •  MDL number MFCD00031687

Purchase

Properties

Related Categories Chemical Synthesis, Phase Transfer Catalysts, Phosphonium Salts, Synthetic Reagents
InChI Key   AKXUUJCMWZFYMV-UHFFFAOYSA-M
grade   technical
concentration   ~80% in H2O

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Signal word 
Danger
Hazard statements 
RIDADR 
UN2810 - class 6.1 - PG 3 - EHS - Toxic, liquids, organic, n.o.s., HI: all
WGK Germany 
3
RTECS 
TA2450000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

404861 Tetrakis(hydroxymethyl)phosphonium chloride solution, 80% in H2O

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?