• USA Home
  • 87648 - (R)-(−)-1,2,3,4-Tetrahydro-1-naphthol

EMAIL THIS PAGE TO A FRIEND
87648 Sigma-Aldrich

(R)-(−)-1,2,3,4-Tetrahydro-1-naphthol

puriss., ≥99.0% (sum of enantiomers, GC)

Synonym: (R)-(−)-α-Tetralol

  • CAS Number 23357-45-1

  • Empirical Formula (Hill Notation) C10H12O

  • Molecular Weight 148.20

  •  Beilstein Registry Number 4350913

  •  MDL number MFCD00063005

  •  PubChem Substance ID 329769590

Purchase

Properties

Related Categories Alcohols, Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Organic Building Blocks More...
InChI Key   JAAJQSRLGAYGKZ-SNVBAGLBSA-N
grade   puriss.
assay   ≥99.0% (sum of enantiomers, GC)
optical activity   [α]20/D −33±1°, c = 2.5% in chloroform
optical purity   enantiomeric ratio: 99:1 (GC)
bp   140 °C/17 mmHg(lit.)
mp   37-39 °C
  39-40 °C(lit.)
density   1.09 g/mL at 25 °C(lit.)

Description

Application

Chiral alcohol

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?