• USA Home
  • 87650 - 1,2,3,4-Tetrahydro-1-naphthol

EMAIL THIS PAGE TO A FRIEND
87650 Sigma-Aldrich

1,2,3,4-Tetrahydro-1-naphthol

purum, ≥97.0% (GC)

Synonym: α-Tetralol

  • CAS Number 529-33-9

  • Empirical Formula (Hill Notation) C10H12O

  • Molecular Weight 148.20

  •  Beilstein Registry Number 2046227

  •  EC Number 208-459-0

  •  MDL number MFCD00001739

  •  PubChem Substance ID 24888928

Purchase

Properties

Related Categories Building Blocks, C9 to C20+, Chemical Synthesis, Organic Building Blocks, Oxygen Compounds,
InChI Key   JAAJQSRLGAYGKZ-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (GC)
refractive index   n20/D 1.564(lit.)
bp   102-104 °C/2 mmHg(lit.)
mp   28-32 °C
density   1.09 g/mL at 25 °C(lit.)
storage temp.   2-8°C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
QL5075000
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

122408 1,2,3,4-Tetrahydro-1-naphthol, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?