EMAIL THIS PAGE TO A FRIEND
87661 Sigma-Aldrich

α-Tetralone

purum, ≥96.0% (GC)

Synonym: 3,4-Dihydro-1(2H)-naphthalenone, alpha-Tetralone

  • CAS Number 529-34-0

  • Empirical Formula (Hill Notation) C10H10O

  • Molecular Weight 146.19

  •  Beilstein Registry Number 607374

  •  EC Number 208-460-6

  •  MDL number MFCD00001688

  •  PubChem Substance ID 24888931

Purchase

Properties

Related Categories Building Blocks, C10, Carbonyl Compounds, Chemical Synthesis, Ketones,
InChI Key   XHLHPRDBBAGVEG-UHFFFAOYSA-N
grade   purum
assay   ≥96.0% (GC)
color   brown
refractive index   n20/D 1.568(lit.)
  n20/D 1.569
bp   113-116 °C/6 mmHg(lit.)
mp   2-7 °C(lit.)
density   1.099 g/mL at 25 °C(lit.)
Gene Information   human ... CYP2A6(1548)
mouse ... Cyp2a5(13087)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
QK4375000
Flash Point(F) 
230 °F
Flash Point(C) 
110 °C
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

T19003 α-Tetralone, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?