• USA Home
  • 87690 - N,N,N′,N′-Tetramethylethylenediamine

EMAIL THIS PAGE TO A FRIEND
87690 Sigma-Aldrich

N,N,N′,N′-Tetramethylethylenediamine

purum, ≥98.0% (GC)

Synonym: 1,2-Bis(dimethylamino)ethane, TEMED, TMEDA

  • CAS Number 110-18-9

  • Linear Formula (CH3)2NCH2CH2N(CH3)2

  • Molecular Weight 116.20

  •  Beilstein Registry Number 1732991

  •  EC Number 203-744-6

  •  MDL number MFCD00008335

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Nitrogen Compounds, Organic Building Blocks, Polyamines More...
InChI Key   KWYHDKDOAIKMQN-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
expl. lim.   9.08 %
impurities   ≤1% water
refractive index   n20/D 1.4179(lit.)
  n20/D 1.418
bp   120-122 °C(lit.)
mp   −55 °C(lit.)
density   0.775 g/mL at 20 °C(lit.)

Description

Other Notes

Substitute for the cancerogenic HMPA as additive for reactions with alkynyl lithium reagents in THF

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2372 3 / PGII
WGK Germany 
1
RTECS 
KV7175000
Flash Point(F) 
68 °F
Flash Point(C) 
20 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

T22500 N,N,N′,N′-Tetramethylethylenediamine, ReagentPlus®, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?