• USA Home
  • 87697 - Tetramethylammonium benzoate

EMAIL THIS PAGE TO A FRIEND
87697 Sigma-Aldrich

Tetramethylammonium benzoate

electrochemical grade, ≥98.5% (NT)

  • CAS Number 25255-90-7

  • Linear Formula (CH3)4N(C6H5COO)

  • Molecular Weight 195.26

  •  Beilstein Registry Number 4925215

  •  MDL number MFCD00270145

  •  PubChem Substance ID 329769597

Purchase

Properties

Related Categories Ammonium Salts, Analytical Reagents, Analytical/Chromatography, Chemical Synthesis, Electrochemistry,
grade   electrochemical grade
InChI Key   IEVVGBFMAHJELO-UHFFFAOYSA-M
assay   ≥98.5% (NT)
mp   188-192 °C (dec.)
solubility   H2O: soluble0.1 g/mL, clear, colorless

Description

General description

Visit our Sensor Applications portal to learn more.

Other Notes

Sales restrictions may apply

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?