• USA Home
  • 87720 - Tetramethylammonium chloride

EMAIL THIS PAGE TO A FRIEND
87720 Sigma-Aldrich

Tetramethylammonium chloride

purum, ≥98.0% (AT)

Synonym: TMA

  • CAS Number 75-57-0

  • Linear Formula (CH3)4N(Cl)

  • Molecular Weight 109.60

  •  Beilstein Registry Number 3911201

  •  EC Number 200-880-8

  •  MDL number MFCD00011628

  •  PubChem Substance ID 329769601

Purchase

Properties

Related Categories Ammonium Salts, Chemical Synthesis, Essential Chemicals, Phase Transfer Catalysts, Research Essentials,
InChI Key   OKIZCWYLBDKLSU-UHFFFAOYSA-M
grade   purum
assay   ≥98.0% (AT)
impurities   ≤2% water
mp   >300 °C(lit.)

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Target organs 
Central nervous system
RIDADR 
UN 2811 6.1 / PGII
WGK Germany 
1
RTECS 
BS7700000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?