• USA Home
  • 87744 - Tetramethylammonium tribromide

EMAIL THIS PAGE TO A FRIEND
87744 Sigma-Aldrich

Tetramethylammonium tribromide

purum, ≥98.0% (AT)

Synonym: Tetramethylammonium bromide perbromide

Purchase

Properties

Related Categories Ammonium Salts, Chemical Synthesis, Phase Transfer Catalysts, Synthetic Reagents
InChI Key   JFZBJMYWDMJRDB-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (AT)
mp   115-118 °C

Description

Other Notes

Mild brominating agent

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3263 8 / PGIII
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?