• USA Home
  • 87747 - Tetramethylammonium tetrafluoroborate

EMAIL THIS PAGE TO A FRIEND
87747 Sigma-Aldrich

Tetramethylammonium tetrafluoroborate

purum, ≥98.0% (T)

  • CAS Number 661-36-9

  • Linear Formula (CH3)4N(BF4)

  • Molecular Weight 160.95

  •  Beilstein Registry Number 3715404

  •  EC Number 211-547-1

  •  MDL number MFCD00011745

  •  PubChem Substance ID 329769605

Purchase

Properties

Related Categories Ammonium Salts, Chemical Synthesis, Phase Transfer Catalysts, Synthetic Reagents
InChI Key   XWFABLFRYCHILB-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (T)
mp   ≥300 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
BS8300000

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?