• USA Home
  • 87765 - 1,1,3,3-Tetramethylbutyl isocyanide

EMAIL THIS PAGE TO A FRIEND
87765 Sigma-Aldrich

1,1,3,3-Tetramethylbutyl isocyanide

purum, ≥95.0% (GC)

Synonym: tert-Octyl isocyanide, Walborsky’s reagent

  • CAS Number 14542-93-9

  • Linear Formula (CH3)3CCH2C(CH3)2NC

  • Molecular Weight 139.24

  •  Beilstein Registry Number 2076559

  •  EC Number 238-577-8

  •  MDL number MFCD00000003

  •  PubChem Substance ID 24888960

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Isocyanides, Nitrogen Compounds, Organic Building Blocks More...
InChI Key   YVPXQMYCTGCWBE-UHFFFAOYSA-N
grade   purum
assay   ≥95.0% (GC)
refractive index   n20/D 1.422(lit.)
  n20/D 1.422
bp   55-57 °C/11 mmHg(lit.)
density   0.794 g/mL at 25 °C(lit.)
storage temp.   2-8°C

Description

Other Notes

Reaction with organometallic reagents, a review; Synthesis of ketones

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 1993C 3 / PGIII
WGK Germany 
3
Flash Point(F) 
125.6 °F
Flash Point(C) 
52 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

226491 1,1,3,3-Tetramethylbutyl isocyanide, technical grade

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?