• USA Home
  • 87835 - 1,1,3,3-Tetramethyldisiloxane

EMAIL THIS PAGE TO A FRIEND
87835 Sigma-Aldrich

1,1,3,3-Tetramethyldisiloxane

purum, ≥97.0% (GC)

Purchase

Properties

Related Categories Chemical Synthesis, Organometallic Reagents, Organosilicon, Siloxanes
InChI Key   NVYQDQZEMGUESH-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (GC)
refractive index   n20/D 1.370(lit.)
  n20/D 1.370
bp   70-71 °C(lit.)
density   0.76 g/mL at 25 °C(lit.)

Description

Other Notes

Reducing agent

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS02  GHS02
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1993BF 3 / PGII
WGK Germany 
2
RTECS 
JN1000000
Flash Point(F) 
-14.8 °F
Flash Point(C) 
-26 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

235733 1,1,3,3-Tetramethyldisiloxane, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?