• USA Home
  • 87925 - Tetramethyltetraselenafulvalene

EMAIL THIS PAGE TO A FRIEND
87925 Sigma-Aldrich

Tetramethyltetraselenafulvalene

purum

Synonym: TMTSF

  • CAS Number 55259-49-9

  • Empirical Formula (Hill Notation) C10H12Se4

  • Molecular Weight 448.04

  •  Beilstein Registry Number 1619644

  •  MDL number MFCD00011507

  •  PubChem Substance ID 24889008

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Organic Building Blocks, Selenium Compounds
InChI Key   YMWLPMGFZYFLRP-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (CHN)
mp   273-275 °C (dec.)(lit.)
storage temp.   2-8°C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN3283 - class 6.1 - PG 3 - EHS - Selenium compound, solid, n.o.s., HI: all (not BR)
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?