• USA Home
  • 87985 - Tetraoctadecylammonium bromide

EMAIL THIS PAGE TO A FRIEND
87985 Sigma-Aldrich

Tetraoctadecylammonium bromide

Selectophore

Purchase

Properties

Related Categories Analytical/Chromatography, Ion Sensor Materials, Ionophores Potentiometric for ISE More...
grade   Selectophore
InChI Key   BUXYZRAZWWVKDS-UHFFFAOYSA-M
mp   103-105 °C(lit.)
  103-105 °C

Description

General description

Visit our Sensor Applications portal to learn more.

Other Notes

Potentiometric properties in PVC membranes ;

Legal Information

Selectophore is a trademark of Sigma-Aldrich Chemie GmbH

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

91664 Tridodecylmethylammonium nitrate, Selectophore, ≥99.0%
72549 Nitrate ionophore - cocktail A, Selectophore
87983 Tetraoctylammonium nitrate, Selectophore, ≥99.0%
07295 Nitrate Ionophore VI, Selectophore, function tested

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?