• USA Home
  • 87990 - Tetraoctadecylammonium bromide

EMAIL THIS PAGE TO A FRIEND
87990 Sigma-Aldrich

Tetraoctadecylammonium bromide

purum, ≥98.0% (NT)

  • CAS Number 63462-99-7

  • Linear Formula [CH3(CH2)17]4N(Br)

  • Molecular Weight 1107.86

  •  Beilstein Registry Number 5846481

  •  EC Number 264-211-1

  •  MDL number MFCD00043172

  •  PubChem Substance ID 24889025

Purchase

Properties

Related Categories Ammonium Salts, Chemical Synthesis, Phase Transfer Catalysts, Synthetic Reagents
InChI Key   BUXYZRAZWWVKDS-UHFFFAOYSA-M
grade   purum
assay   ≥98.0% (NT)
mp   103-105 °C(lit.)
  103-105 °C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?