• USA Home
  • 88000 - Tetraoctylammonium bromide

EMAIL THIS PAGE TO A FRIEND
88000 Sigma-Aldrich

Tetraoctylammonium bromide

purum, ≥98.0% (AT)

Purchase

Properties

Related Categories Ammonium, Ammonium Salts, Chemical Synthesis, Ionic Liquids, Phase Transfer Catalysts,
InChI Key   QBVXKDJEZKEASM-UHFFFAOYSA-M
grade   purum
assay   ≥98.0% (AT)
mp   95-98 °C(lit.)
  96-99 °C

Description

Other Notes

PTC catalyst for preparing alkynes from dichlorides or chloroalkenes

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

294136 Tetraoctylammonium bromide, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?