• USA Home
  • 88065 - Tetraphenylphosphonium tetraphenylborate

EMAIL THIS PAGE TO A FRIEND
88065 Sigma-Aldrich

Tetraphenylphosphonium tetraphenylborate

Selectophore

Synonym: Tetraphenylboron tetraphenylphosphonium

  • CAS Number 15525-15-2

  • Empirical Formula (Hill Notation) C48H40BP

  • Molecular Weight 658.62

  •  Beilstein Registry Number 3890798

  •  MDL number MFCD00145389

  •  PubChem Substance ID 329769629

Purchase

Properties

Related Categories Additives for ISE, Analytical/Chromatography, Ion Sensor Materials More...
grade   Selectophore
InChI Key   QYFWPOPFFBCTLK-UHFFFAOYSA-N

Description

General description

Visit our Sensor Applications portal to learn more.

Other Notes

Lipophilic salt used in liquid-membrane anion-selective electrodes

Legal Information

Selectophore is a trademark of Sigma-Aldrich Chemie GmbH

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?