• USA Home
  • 88105 - Tetrapropylammonium bromide

EMAIL THIS PAGE TO A FRIEND
88105 Sigma-Aldrich

Tetrapropylammonium bromide

purum, ≥98.0% (AT)

  • CAS Number 1941-30-6

  • Linear Formula (CH3CH2CH2)4N(Br)

  • Molecular Weight 266.26

  •  Beilstein Registry Number 3567846

  •  EC Number 217-727-6

  •  MDL number MFCD00011840

Purchase

Properties

Related Categories Ammonium Salts, Chemical Synthesis, Phase Transfer Catalysts, Synthetic Reagents
InChI Key   BGQMOFGZRJUORO-UHFFFAOYSA-M
grade   purum
assay   ≥98.0% (AT)
solubility   H2O: soluble0.1 g/mL, clear

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

225568 Tetrapropylammonium bromide, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?