• USA Home
  • 88106 - Tetrapropylammonium bisulfate

EMAIL THIS PAGE TO A FRIEND
88106 Sigma-Aldrich

Tetrapropylammonium bisulfate

for ion pair chromatography, ≥99.0%

Synonym: Tetrapropylammonium hydrogen sulfate

  • CAS Number 56211-70-2

  • Empirical Formula (Hill Notation) C12H29NO4S

  • Molecular Weight 283.43

  •  Beilstein Registry Number 6833419

  •  MDL number MFCD00038764

  •  PubChem Substance ID 329769634

Purchase

Properties

Related Categories Acidic Ion Pair Reagents (Anionic), Analytical Reagents, Analytical/Chromatography, Chromatography Reagents & Solvents, Ion Pair Reagents More...
grade   for ion pair chromatography
InChI Key   MOXJKKOSZCHGEU-UHFFFAOYSA-M
assay   ≥99.0% (T)
  ≥99.0%
mp   158-160 °C
λ   10 % in H2O
UV absorption   λ: 210 nm Amax: 0.05
  λ: 220 nm Amax: 0.04
  λ: 230 nm Amax: 0.03
  λ: 260 nm Amax: 0.02
  λ: 500 nm Amax: 0.02
suitability   corresponds to standard for RP gradient test
  corresponds to standard for filter test

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?