• USA Home
  • 88110 - Tetrapropylammonium hydroxide solution

EMAIL THIS PAGE TO A FRIEND
88110 Sigma-Aldrich

Tetrapropylammonium hydroxide solution

technical, ~20% in H2O (~1.0 M)

  • CAS Number 4499-86-9

  • Linear Formula (CH3CH2CH2)4N(OH)

  • Molecular Weight 203.36

  •  Beilstein Registry Number 4207983

  •  EC Number 224-800-6

  •  MDL number MFCD00009360

Purchase

Properties

Related Categories Acids & Bases, Bases, Chemical Synthesis, Organic Bases, Synthetic Reagents More...
InChI Key   LPSKDVINWQNWFE-UHFFFAOYSA-M
grade   technical
concentration   ~20% in H2O (~1.0 M)
impurities   ≤3% halides (as bromide)
density   1.00 g/mL at 20 °C
anion traces   sulfate (SO42-): ≤1000 mg/kg

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3267 8 / PGII
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Related Content

Molarity Calculator and Normality Calculator for Acids and Bases

The molarity calculator tool provides lab-ready directions describing how to prepare an acid or base solution of specified Molarity (M) or Normality (N) from a concentrated acid or base solution.  To...

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?