EMAIL THIS PAGE TO A FRIEND
88143 Sigma-Aldrich

Tetratetracontane

purum, ≥95.0% (GC)

Purchase

Properties

Related Categories Acyclic, Alkanes, Building Blocks, Chemical Synthesis, Organic Building Blocks More...
InChI Key   KMXFZRSJMDYPPG-UHFFFAOYSA-N
grade   purum
assay   ≥95.0% (GC)
mp   85-87 °C(lit.)
  85-87 °C

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?