EMAIL THIS PAGE TO A FRIEND
88272 Sigma-Aldrich

Thallium(I) nitrate

purum p.a., ≥99.0% (T)

Purchase

Properties

Related Categories Materials Science, Micro/NanoElectronics, Solution Deposition Precursors, Thallium
InChI Key   FYWSTUCDSVYLPV-UHFFFAOYSA-N
grade   purum p.a.
assay   ≥99.0% (T)
form   solid
mp   206 °C(lit.)
anion traces   chloride (Cl-): ≤50 mg/kg
cation traces   Ca: ≤50 mg/kg
  Cd: ≤50 mg/kg
  Co: ≤50 mg/kg
  Cu: ≤50 mg/kg
  Fe: ≤50 mg/kg
  K: ≤100 mg/kg
  Na: ≤100 mg/kg
  Ni: ≤50 mg/kg
  Pb: ≤300 mg/kg
  Zn: ≤50 mg/kg

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Signal word 
Danger
Hazard statements 
RIDADR 
UN 2727 5.1(6.1) / PGII
WGK Germany 
2
RTECS 
XG5950000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?