• USA Home
  • 88302 - 2-Thenoyltrifluoroacetone

EMAIL THIS PAGE TO A FRIEND
88302 Sigma-Aldrich

2-Thenoyltrifluoroacetone

purum, ≥97.0% (GC)

Synonym: 4,4,4-Trifluoro-1-(2-thienyl)-1,3-butanedione, TTA (extractant), TTB, TTFA

  • CAS Number 326-91-0

  • Empirical Formula (Hill Notation) C8H5F3O2S

  • Molecular Weight 222.18

  •  Beilstein Registry Number 168645

  •  EC Number 206-316-7

  •  MDL number MFCD00005445

  •  PubChem Substance ID 329769644

Purchase

Properties

Related Categories Building Blocks, C7 to C9, Chemical Synthesis, Heterocyclic Building Blocks, Thiophenes More...
InChI Key   TXBBUSUXYMIVOS-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (GC)
bp   96-98 °C/8 mmHg(lit.)
mp   40-44 °C(lit.)
  40-44 °C
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
233.6 °F
Flash Point(C) 
112 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?