EMAIL THIS PAGE TO A FRIEND
88304 Sigma-Aldrich

Theobromine

≥98.0% (HPLC)

Synonym: 2,6-Dihydroxy-3,7-dimethylpurine, 3,7-Dihydro-3,7-dimethyl-1H-purine-2,6-dione, 3,7-Dimethylxanthine

  • CAS Number 83-67-0

  • Empirical Formula (Hill Notation) C7H8N4O2

  • Molecular Weight 180.16

  •  Beilstein Registry Number 16464

  •  EC Number 201-494-2

  •  MDL number MFCD00022830

Purchase

Properties

Related Categories Alkaloid, Building Blocks, Cell Biology, Chemical Synthesis, Heterocyclic Building Blocks,
InChI Key   YAPQBXQYLJRXSA-UHFFFAOYSA-N
assay   ≥98.0% (HPLC)
mp   345-350 °C

Description

Biochem/physiol Actions

Phosphosdiesterase inhibitor; weak adenosine receptor antagonist; diuretic; smooth muscle relaxant.

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.




All labs need water
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
XH2275000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

T4500 Theobromine, ≥98.0%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?