• USA Home
  • 88330 - Chloro(dimethyl)thexylsilane

EMAIL THIS PAGE TO A FRIEND
88330 Sigma-Aldrich

Chloro(dimethyl)thexylsilane

purum, ≥97.0% (GC)

Synonym: Chlorodimethyl(1,1,2-trimethylpropyl)silane, Dimethyl-(2,3-dimethyl-2-butyl)chlorosilane, Dimethyl-thexylchlorosilane, Dimethylthexylsilyl chloride, TDSCl, Thexyldimethylsilyl chloride

  • CAS Number 67373-56-2

  • Linear Formula (CH3)2CHC(CH3)2Si(CH3)2Cl

  • Molecular Weight 178.77

  •  Beilstein Registry Number 3647960

  •  MDL number MFCD00012175

  •  PubChem Substance ID 329769647

Purchase

Properties

Related Categories Chemical Synthesis, Protecting and Derivatizing Reagents, Protection and Derivatization, Silicon-Based, Synthetic Reagents More...
InChI Key   KIGALSBMRYYLFJ-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (GC)
refractive index   n20/D 1.449(lit.)
  n20/D 1.450
bp   55-56 °C/10 mmHg(lit.)
density   0.909 g/mL at 25 °C(lit.)

Description

Other Notes

Protecting group reagent with some advantages compared to tert-butyldimethylchlorosilane (less expensive, liquid, slightly more stable)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2986 3(8) / PGII
WGK Germany 
3
Flash Point(F) 
114.8 °F
Flash Point(C) 
46 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?