• USA Home
  • 88425 - 3-(2-Thienyl)-DL-alanine

EMAIL THIS PAGE TO A FRIEND
88425 Sigma-Aldrich

3-(2-Thienyl)-DL-alanine

purum, ≥98.0% (NT)

Synonym: 2-Amino-3-(2-thienyl)propionic acid

  • CAS Number 2021-58-1

  • Empirical Formula (Hill Notation) C7H9NO2S

  • Molecular Weight 171.22

  •  Beilstein Registry Number 82874

  •  EC Number 217-967-1

  •  MDL number MFCD00005459

  •  PubChem Substance ID 329769651

Purchase

Properties

Related Categories Alanine Derivatives, Chemical Biology, Chemical Synthesis, Peptide Chemistry, Unnatural Amino Acids & Derivatives More...
InChI Key   WTOFYLAWDLQMBZ-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (NT)
mp   ~270 °C (dec.)
  275-277 °C (dec.)(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

287288 3-(2-Thienyl)-DL-alanine, ≥98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?