• USA Home
  • 88439 - Nickel chloride 0.01M – TRIS-HCl pH 8.5; 0.1M – PEG MME 2000 20% (w/v) solution

EMAIL THIS PAGE TO A FRIEND
88439 Sigma-Aldrich

Nickel chloride 0.01M – TRIS-HCl pH 8.5; 0.1M – PEG MME 2000 20% (w/v) solution

Synonym: Crystallization solution 45/Kit-No 70437

Purchase

Properties

pH   8.5±0.2 ( in neat)
density   1.04 g/mL at 20 °C
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
WGK Germany 
2

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?