88450 Sigma-Aldrich

Thioacetamide

ACS reagent, for the precipitation (of heavy metals), ≥99.0%

DOWNLOAD MSDS (PDF)

Synonym: Ethanethioamide

  • CAS Number 62-55-5

  • Linear Formula CH3CSNH2

  • Molecular Weight 75.13

  •  Beilstein Registry Number 506006

  •  EC Number 200-541-4

  •  MDL number MFCD00008070

  •  PubChem Substance ID 24889097

Properties

Related Categories Analytical Reagents, Analytical Reagents for General Use, Analytical/Chromatography, Gravimetric Reagents, Puriss p.a. ACS,
grade   ACS reagent
  for the precipitation (of heavy metals)
assay   ≥99.0% (RT)
  ≥99.0%
ign. residue   ≤0.05% (as SO4)
mp   108-112 °C(lit.)
  111-114 °C
cation traces   Ca: ≤10 mg/kg
  Cd: ≤5 mg/kg
  Co: ≤5 mg/kg
  Cr: ≤5 mg/kg
  Cu: ≤5 mg/kg
  Fe: ≤5 mg/kg
  K: ≤50 mg/kg
  Mg: ≤5 mg/kg
  Mn: ≤5 mg/kg
  Na: ≤100 mg/kg
  Ni: ≤5 mg/kg
  Pb: ≤5 mg/kg
  Zn: ≤5 mg/kg

Description

Other Notes

Used e.g. for the decomposition of amino acid copper complexes1

Price and Availability

Customers Also Viewed

Glycerin

meets USP testing specifications

Lead(II) nitrate

puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.0%

Thioacetamide

ACS reagent, ≥99.0%

Thioacetamide

reagent grade, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?