• USA Home
  • 88481 - 2-Thiobarbituric acid

EMAIL THIS PAGE TO A FRIEND
88481 Sigma-Aldrich

2-Thiobarbituric acid

purum, ≥98.0% (S)

Synonym: 4,6-Dihydroxy-2-mercaptopyrimidine, 4,6-Dihydroxypyrimidine-2-thiol, TBA

  • CAS Number 504-17-6

  • Empirical Formula (Hill Notation) C4H4N2O2S

  • Molecular Weight 144.15

  •  Beilstein Registry Number 120663

  •  EC Number 207-985-8

  •  MDL number MFCD00006674

  •  PubChem Substance ID 24889103

Purchase

Properties

Related Categories Building Blocks, C4 to C5, Chemical Synthesis, Heterocyclic Building Blocks, Pyrimidines More...
InChI Key   RVBUGGBMJDPOST-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (S)
drug control   regulated under CDSA - not available from Sigma-Aldrich Canada
ign. residue   ≤0.1%
mp   245 °C (dec.)(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

RIDADR 
UN 3335
WGK Germany 
2
RTECS 
CQ7700000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

T5500 2-Thiobarbituric acid, ≥98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?