EMAIL THIS PAGE TO A FRIEND
88491 Sigma-Aldrich

Thiobenzamide

purum, ≥98.0% (N)

  • CAS Number 2227-79-4

  • Linear Formula C6H5CSNH2

  • Molecular Weight 137.20

  •  Beilstein Registry Number 606021

  •  EC Number 218-765-6

  •  MDL number MFCD00008060

Purchase

Properties

InChI Key   QIOZLISABUUKJY-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (N)
color   yellow
mp   113-117 °C(lit.)
  113-117 °C

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2811 6.1 / PGIII
WGK Germany 
3
RTECS 
CV5860000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

148229 Thiobenzamide, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?