EMAIL THIS PAGE TO A FRIEND
88500 Sigma-Aldrich

Thiobenzoic acid

technical, ≥90% (T)

  • CAS Number 98-91-9

  • Linear Formula C6H5COSH

  • Molecular Weight 138.19

  •  Beilstein Registry Number 2039640

  •  EC Number 202-712-9

  •  MDL number MFCD00004852

  •  PubChem Substance ID 24889105

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Organic Building Blocks, Sulfur Compounds, Thiocarbonyl Compounds More...
InChI Key   UIJGNTRUPZPVNG-UHFFFAOYSA-N
grade   technical
assay   ≥90% (T)
refractive index   n20/D 1.605(lit.)
  n20/D 1.605
bp   122 °C/30 mmHg(lit.)
mp   15-18 °C(lit.)
density   1.174 g/mL at 25 °C(lit.)
storage temp.   2-8°C

Description

Other Notes

This product has been replaced by T28207-ALDRICH | Thiobenzoic acid technical grade, 90%

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 2810 6.1 / PGIII
WGK Germany 
3
RTECS 
DH6839000
Flash Point(F) 
201.2 °F
Flash Point(C) 
94 °C
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

T28207 Thiobenzoic acid, technical grade, 90%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?